missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VRK2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93930-0.1ml
This item is not returnable.
View return policy
Description
VRK2 Polyclonal antibody specifically detects VRK2 in Human samples. It is validated for Western Blot
Specifications
| VRK2 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| EC 2.7.11.1, serine/threonine-protein kinase VRK2, vaccinia related kinase 2, vaccinia virus B1R-related kinase 2, Vaccinia-related kinase 2, vaccinia-related kinase-2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 269-508 of human VRK2 (NP_006287.2). DELPQSVLKWAPSGSSCCEIAQFLVCAHSLAYDEKPNYQALKKILNPHGIPLGPLDFSTKGQSINVHTPNSQKVDSQKAATKQVNKAHNRLIEKKVHSERSAESCATWKVQKEEKLIGLMNNEAAQESTRRRQKYQESQEPLNEVNSFPQKISYTQFPNSFYEPHQDFTSPDIFKKSRSPSWYKYTSTVSTGITDLESSTGLWPTISQFTLSEETNADVYYYRIIIPVLLMLVFLALFFL | |
| 0.1 mL | |
| Protein Kinase | |
| 7444 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction