missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VRK3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15938744
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15938744 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15938744 Supplier Invitrogen™ Supplier No. PA566016

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: CPYCGNSLPVE EHVGSQTFVN PHVSSFQGSK RGLNSSFETS PKKVKWSSTV TSPRLSLFSD GDSSESEDTL SS Highest antigen sequence identity to the following orthologs - mouse 68%, rat 73%.

This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VRK3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene VRK3
Gene Accession No. Q8IV63
Gene Alias Inactive serine/threonine-protein kinase VRK3; serine/threonine-protein kinase VRK3; serine/threonine-protein pseudokinase VRK3; vaccinia related kinase 3; vaccinia-related kinase 3; VRK3
Gene Symbols VRK3
Host Species Rabbit
Immunogen Recombinant Human VRK3. Recombinant protein control fragment (Product #RP-106702).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51231
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.