missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VRK3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15608-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a member of the vaccinia-related kinase family of serine/threonine protein kinases. In both human and mouse, this gene has substitutions at several residues within the ATP binding motifs that in other kinases have been shown to be required for catalysis. In vitro assays indicate the protein lacks phosphorylation activity. The protein, however, likely retains its substrate binding capability. This gene is widely expressed in human tissues and its protein localizes to the nucleus. Alternative splicing results in multiple transcripts encoding different isoforms.Specifications
| VRK3 | |
| Polyclonal | |
| Unconjugated | |
| VRK3 | |
| vaccinia related kinase 3, VRK3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51231 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8IV63 | |
| VRK3 | |
| VRK3 Fusion Protein Ag7993 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction