missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VRTN Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551581
This item is not returnable.
View return policy
Description
Immunogen sequence: ATLEAIFDAD VKASCFPSSF SNVWHLYALA SVLQRNIYSI YPMRNLKIRP YFNRVIRPRR CDHVPSTLHI MWAGQPLTSH FFRHQYFAPV VGLEEVEAEG APGVAPALPA LAPLSSPAKT LELLNREPGL SYSHLC Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 82%.
VRTN gene ontology annotations related to this gene include transposition, DNA-mediated.
Specifications
| VRTN | |
| Polyclonal | |
| Unconjugated | |
| VRTN | |
| 7420416P09Rik; C14orf115; vertebrae development associated; vertebrae development homolog (pig); vertnin; Vrtn | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55237 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9H8Y1 | |
| VRTN | |
| Recombinant protein corresponding to Human VRTN. Recombinant protein control fragment (Product #RP-88860). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction