missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA584527
This item is not returnable.
View return policy
Description
Immunogen sequence: LTCQVSGAYP PAKILWLRNL TQPEVIIQPS SRHLITQDGQ NSTLTIHNCS QDLDEGYYIC RADSPVGVRE MEIWLSVK Highest antigen sequence indentity to the following orthologs: Rat - 74%, Mouse - 72%.
VSIG10 (V-Set And Immunoglobulin Domain Containing 10) is a Protein Coding gene. Diseases associated with VSIG10 include Adenoid Hypertrophy. An important paralog of this gene is CNTN5. [GeneCards].
Specifications
| VSIG10 | |
| Polyclonal | |
| Unconjugated | |
| VSIG10 | |
| RGD1565800; V-set and immunoglobulin domain containing 10; V-set and immunoglobulin domain-containing protein 10; Vsig10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N0Z9 | |
| VSIG10 | |
| Recombinant protein corresponding to Human VSIG10. Recombinant protein control fragment (Product #RP-108464). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction