missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG3/IGSF11 Antibody (973408), mFluor Violet 450 SE, Novus Biologicals™
Shop All Bio Techne ProductsDescription
VSIG3/IGSF11 Monoclonal antibody specifically detects VSIG3/IGSF11 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
Specifications
| Conjugate | mFluor Violet 450 SE |
| Dilution | Flow Cytometry, CyTOF-ready |
| Immunogen | Mouse myeloma cell line NS0-derived recombinant human VSIG3, Met1-Gly245, Accession # Q5DX21 |
| Primary or Secondary | Primary |
| Target Species | Human |
| Isotype | IgG1 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?