missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG3/IGSF11 Antibody (973422) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB92293JF525
This item is not returnable.
View return policy
Description
VSIG3/IGSF11 Monoclonal antibody specifically detects VSIG3/IGSF11 in Human samples. It is validated for Flow Cytometry,Blockade of Receptor-ligand Interaction,CyTOF
Specifications
| VSIG3/IGSF11 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein | |
| Mouse myeloma cell line NS0-derived recombinant human VSIG3, Met1-Gly245, Accession # Q5DX21 | |
| RUO | |
| 152404 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| 973422 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction