missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG3/IGSF11 Antibody (OTI2H1), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74861V
This item is not returnable.
View return policy
Description
VSIG3/IGSF11 Monoclonal specifically detects VSIG3/IGSF11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| VSIG3/IGSF11 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| IGSF11 | |
| Recombinant protein fragment corresponding to amino acids 23-241 of human VSIG3/IGSF11 (NP_689751) produced in E.coli. | |
| 0.1 mL | |
| 152404.0 | |
| Store at 4°C in the dark. |
| Western Blot | |
| OTI2H1 | |
| Western Blot | |
| Brain and testis-specific immunoglobulin superfamily protein, Bt-IGSF, BTIGSF, cancer/testis antigen 119, CT119, CXADR like 1, CXADRL1, IgSF11, Igsf13, immunoglobulin superfamily member 11, immunoglobulin superfamily, member 11, MGC35227, V-set and immunoglobulin domain containing 3, V-set and immunoglobulin domain-containing protein 3, VSIG3brain and testis-specific immunoglobin superfamily protein | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction