missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Antibody (2174B), PE/Cy5.5, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB46461PECY55
This item is not returnable.
View return policy
Description
VSIG4 Monoclonal antibody specifically detects VSIG4 in Human samples. It is validated for Flow Cytometry
Specifications
| VSIG4 | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 11326 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG |
| Flow Cytometry | |
| 2174B | |
| Flow Cytometry | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Mouse myeloma cell line NS0-derived human VSIG4, Arg20-Pro283, Accession # Q9Y279 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction