missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 136.00 - € 321.00
Specifications
| Antigen | VSIG4 |
|---|---|
| Dilution | ELISA 1:5000-1:10000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18619865
|
Novus Biologicals
NBP2-99947-20ul |
20 μg |
€ 136.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18603586
|
Novus Biologicals
NBP2-99947-100ul |
100 μL |
€ 321.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIG4 Polyclonal antibody specifically detects VSIG4 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| VSIG4 | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Produced in rabbits immunized with purified, recombinant Human VSIG4 (Q9Y279-1; Met1-Pro283). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| ELISA 1:5000-1:10000, Immunohistochemistry 1:500-1:2000, Immunohistochemistry-Paraffin 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 11326 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title