missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VSIG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA542027
This item is not returnable.
View return policy
Description
Peptide sequence: VPGDVSLQLS TLEMDDRSHY TCEVTWQTPD GNQVVRDKIT ELRVQKLSVS Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 92%; Horse: 91%; Human: 100%; Mouse: 92%; Pig: 100%; Rabbit: 82%; Rat: 86%.
T cell activation by APCs is positively and negatively regulated by members of the B7 family. VSIG4 is a strong negative regulator of murine and human T cell proliferation and IL-2 production.
Specifications
| VSIG4 | |
| Polyclonal | |
| Unconjugated | |
| VSIG4 | |
| A530061A11; BC025105; complement receptor of the immunoglobulin superfamily; CRIg; Ig superfamily protein; MGC137243 protein; protein Z39Ig; UNQ317/PRO362; V-set and immunoglobulin domain containing 4; V-set and immunoglobulin domain-containing protein 4; VSIG4; Z39IG | |
| Rabbit | |
| Protein A | |
| RUO | |
| 11326 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9Y279 | |
| VSIG4 | |
| Synthetic peptide directed towards the N-terminal of human VSIG4 (aa 92-141). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction