missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG4 Rabbit anti-Human, Clone: 5, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 398.00 - € 510.00
Specifications
| Antigen | VSIG4 |
|---|---|
| Clone | 5 |
| Dilution | ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18210218
|
Novus Biologicals
NBP2-90065-50UL |
50 μg |
€ 398.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18211916
|
Novus Biologicals
NBP2-90065-100UL |
100 μg |
€ 510.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
VSIG4 Monoclonal specifically detects VSIG4 in Human samples. It is validated for ELISA, Sandwich ELISA Detection.Specifications
| VSIG4 | |
| ELISA 1:5000-1:10000, Sandwich ELISA Detection 1:1000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 11326 | |
| IgG | |
| Protein A purified |
| 5 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CRIg, Ig superfamily protein, Protein Z39Ig, V-set and immunoglobulin domain containing 4, V-set and immunoglobulin domain-containing protein 4, Z39IGcomplement receptor of the immunoglobulin superfamily | |
| Recombinant Human VSIG4 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title