missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSIG8 Antibody (961823) [Janelia Fluor™ 549], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9418I
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal antibody specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry, ELISA
Specifications
| VSIG8 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 391123 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA | |
| 961823 | |
| Flow Cytometry, ELISA | |
| V-set and immunoglobulin domain containing 8 | |
| Human embryonic kidney cell line HEK293-derived transfected with human VSIG8, Met1-Gly263, Accession # Q5VU13 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction