missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ VSIG8 Antibody (961829) [PE/Cy7], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ FAB94182PECY7
This item is not returnable.
View return policy
Description
VSIG8 Monoclonal antibody specifically detects VSIG8 in Human samples. It is validated for Flow Cytometry
Specifications
| VSIG8 | |
| Monoclonal | |
| PE-Cyanine7 | |
| V-set and immunoglobulin domain containing 8 | |
| Human embryonic kidney cell line HEK293-derived recombinant human VSIG8, Met1-Gly263, Accession # Q5VU13 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 961829 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 391123 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction