missing translation for 'onlineSavingsMsg'
Learn More

VSNL1 Monoclonal Antibody (OTI2B4), TrueMAB™, OriGene

Product Code. p-200018098
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17510333 30 μL 30µL
17590323 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17510333 Supplier Origene Technologies Supplier No. TA801901S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

VSNL1 Monoclonal antibody specifically detects VSNL1 in Human samples. It is validated for Western Blot

Specifications

Antigen VSNL1
Applications Western Blot
Classification Monoclonal
Clone OTI2B4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene VSNL1
Gene Accession No. P62760
Gene Alias 21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7447
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.