missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSNL1 Monoclonal Antibody (OTI6A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA802024
This item is not returnable.
View return policy
Description
This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined.Specifications
| VSNL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62760 | |
| VSNL1 | |
| Human recombit protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI6A5 | |
| Unconjugated | |
| VSNL1 | |
| HLP3, HPCAL3, HUVISL1, VILIP, VILIP-1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 7447 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction