missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSNL1 Monoclonal Antibody (OTI6B11), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801989S
This item is not returnable.
View return policy
Description
VSNL1 Monoclonal antibody specifically detects VSNL1 in Human samples. It is validated for Western BlotSpecifications
| VSNL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P62760 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7447 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI6B11 | |
| Unconjugated | |
| VSNL1 | |
| 21 kDa CABP; hippocalcin-like protein 3; HLP3; HPCAL3; HUVISL1; Neural visinin-like protein 1; neurocalcin alpha; Neurocalcin-alpha; NVL-1; Nvp1; NVP-1; Ratnvp1; VILIP; VILIP-1; visinin like 1; visinin-like 1; visinin-like protein 1; Visl1; VLP-1; Vnsl1; VSNL1 | |
| Human recombinant protein fragment corresponding to amino acids 2-191 of human VSNL1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction