missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM2A Antibody (1024735) [PerCP], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB100371C
This item is not returnable.
View return policy
Description
VSTM2A Monoclonal antibody specifically detects VSTM2A in Human samples. It is validated for Flow Cytometry, ELISA, CyTOF
Specifications
| VSTM2A | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 222008 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Flow Cytometry, ELISA, CyTOF | |
| 1024735 | |
| Flow Cytometry, ELISA, CyTOF-ready | |
| MGC33530, V-set and transmembrane domain containing 2, V-set and transmembrane domain containing 2A, V-set and transmembrane domain-containing protein 2A, VSTM2 | |
| Human embryonic kidney cell HEK293-derived human VSTM2A protein, Ser25-Phe244, Accession # Q8TAG5-2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction