missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VSTM4 Antibody (2694A), DyLight 755, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne FAB10767Z
This item is not returnable.
View return policy
Description
VSTM4 Monoclonal antibody specifically detects VSTM4 in Human samples. It is validated for Flow Cytometry
Specifications
| VSTM4 | |
| Monoclonal | |
| DyLight 755 | |
| FLJ31737, hypothetical protein LOC196740, V-set and transmembrane domain containing 4 | |
| Chinese Hamster Ovary cell line CHO-derived human VSTM4, Leu24-Tyr180, Accession # Q8IW00-1 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| 2694A | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 196740 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction