Learn More
IKA™ VSW 1 Woulff Bottle
Vacuum safety woulff bottle designed for use with a range of rotary evaporators.
Brand:  IKA™ 0020015406
Additional Details : Weight : 0.35380kg
Product Code. 16288501
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
  • For use with the following IKA™ models:
    • RV 3 V, Vacstar Control, RV 3 V-C, Vacstar Digital, RV 3 FLEX, MVP 10 Basic Vacuum Pump, RV 3 with dry ice condenser (coated), RV 8 V, Vc 10 Vacuum Controller, RV 8 V-C, RV 8 FLEX, RV 10 Auto V, RV 10 Auto V-C, RV 10 Auto FLEX
Specifications
Specifications
Vacuum
0.35 kg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers