missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTA1 Antibody (14G10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59420AF750
This item is not returnable.
View return policy
Description
VTA1 Monoclonal antibody specifically detects VTA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| VTA1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| C6orf55DRG1, Dopamine-responsive gene 1 protein, DRG-1, FLJ27228, homolog of mouse SKD1-binding protein 1, LIP5HSPC228, LYST-interacting protein 5, My012, SBP1chromosome 6 open reading frame 55, SKD1-binding protein 1, vacuolar protein sorting-associated protein VTA1 homolog, Vps20-associated 1 homolog (S. cerevisiae) | |
| Recombinant human VTA1 (1-307aa) purified from E. coli | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 51534 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 14G10 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction