missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VTI1A Antibody (OTI1F4), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-74862CL1
This item is not returnable.
View return policy
Description
VTI1A Monoclonal antibody specifically detects VTI1A in Canine,Human,Monkey,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot
Specifications
| VTI1A | |
| Monoclonal | |
| CoraFluor 1 | |
| MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) | |
| Full length human recombinant protein of human VTI1A(NP_660207) produced in HEK293T cell. | |
| 0.1 mL | |
| Neuroscience, Signal Transduction | |
| 143187 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot | |
| OTI1F4 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Canine, Human, Monkey, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction