missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
VTI1A Monoclonal specifically detects VTI1A in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | VTI1A |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Clone | OTI1F4 |
| Conjugate | DyLight 680 |
| Dilution | Western Blot, Immunohistochemistry |
| Formulation | PBS with 0.05% Sodium Azide |
| Gene Alias | MVti1, vesicle transport through interaction with t-SNAREs homolog 1A, vesicle transport through interaction with t-SNAREs homolog 1A (yeast) |
| Gene Symbols | VTI1A |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?