missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VTI1B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560618
This item is not returnable.
View return policy
Description
Immunogen sequence: ASSEHFEKLH EIFRGLHEDL QGVPERLLGT AGTEEKKKLI RDFDEKQQEA NETLAEMEEE LRYAPLSFRN PMMSKLRNYR KDLAKLHREV RSTPLTATPG GR Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
V-SNARE that mediates vesicle transport pathways through interactions with t-SNAREs on the target membrane. These interactions are proposed to mediate aspects of the specificity of vesicle trafficking and to promote fusion of the lipid bilayers. May be concerned with increased secretion of cytokines associated with cellular senescence. Fischer von Mollard G., J. Biol. Chem. 273:2624-2630(1998). Li H.-C., Biochem. Biophys. Res. Commun. 247:70-74(1998).
Specifications
| VTI1B | |
| Polyclonal | |
| Unconjugated | |
| Vti1b | |
| A novel SNARE; AU015348; GES30; HGNC:17793; MVti1b; SNARE; Soluble N-ethylmaleimide-sensitive factor-attachment protein receptor; vesicle transport through interaction with t-SNAREs 1B; vesicle transport through interaction with t-SNAREs 1B homolog; vesicle transport through interaction with t-SNAREs homolog 1B; Vesicle transport v-SNARE protein Vti1-like 1; vesicle-associated soluble NSF attachment protein receptor; v-SNARE; VTI1; VTI1B; VTI1L; VTI1L1; VTI1-LIKE; vti1-rp1; VTI2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10490 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UEU0 | |
| Vti1b | |
| Recombinant protein corresponding to Human VTI1B. Recombinant protein control fragment (Product #RP-100108). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction