missing translation for 'onlineSavingsMsg'
Learn More

VTP-27999 Hydrochloride, MedChemExpress

Product Code. 16414117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16414117 50 mg 50mg
16438218 10 mg 10mg
16427956 100 mg 100mg
16410658 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16414117 Supplier Medchem Express Supplier No. HY76652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress VTP-27999 Hcl is an alkyl amine Renin inhibitor; VTP-27999 is useful for Hypertension and End-Organ Diseases.

VTP-27999 Hcl is an alkyl amine Renin inhibitor

Specifications

Chemical Name or Material VTP-27999 Hydrochloride
Physical Form Solid
CAS 1264191-73-2
Color Off-White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C26H42Cl2N4O5
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : ≥ 100 mg/mL (178.08 mM)
SMILES O=C(NC[C@H](C[C@@H]1COCCC1)NC)N(CCC2)C[C@]2([H])[C@](OCCNC(OC)=O)([H])C3=CC(Cl)=CC=C3.Cl
Molecular Weight (g/mol) 561.54
Formula Weight 561.54
Percent Purity 98.02%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.