Learn More
Tocris Bioscience™ VTP 27999 trifluoroacetate
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
VTP 27999 trifluoroacetate is a highly potent and selective renin inhibitor (IC50 = 0.47 nM). Displays >1000-fold selectivity over related and unrelated off-targets. Blocks renin-stimulated ERK1/2 phosphorylation in vascular smooth muscle cells. Exhibits a different mode of action to aliskiren (Cat. No. 5491).
Specifications
Specifications
| Chemical Name or Material | Methyl N-[2-[(R)-(3-Chlorophenyl)[(3R)-1-[[[(2S)-2-(methylamino)-3-[(3R)-tetrahydro-2H-pyran-3-yl]propyl]amino]carbonyl]-3-piperidinyl]methoxy]ethyl]carbamate trifluoroacetate |
| CAS | 1013937-63-7 |
| Quantity | 10 mg |
| Target | Additional Synthetic Protease Inhibitors |
| Molecular Formula | C26H41ClN4O5.CF3CO2H |
| Formula Weight | 639.1 |
| Purity | >97% |