missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VU 0240551, MedChemExpress
Description
VU 0240551 is a potent neuronal K-Cl cotransporter KCC2 inhibitor (IC50=560 nM) and is selective versus NKCC1. VU 0240551 also inhibits hERG and L-type Ca2+ channels. VU 0240551 attenuates GABA-induced hyperpolarization of P cells, produces a positive shift in the P cell GABA reversal potential and enhances P cell synaptic transmission.
Specifications
Specifications
| Chemical Name or Material | VU 0240551 |
| Physical Form | Solid |
| CAS | 893990-34-6 |
| Color | White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C16H14N4OS2 |
| Solubility Information | DMSO : ≥ 50 mg/mL (146.01 mM) |
| SMILES | O=C(CSC1=NN=C(C2=CC=CC=C2)C=C1)NC3=NC(C)=CS3 |
| Molecular Weight (g/mol) | 342.44 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?