Learn More
Tocris Bioscience™ VU 0409551
GREENER_CHOICE
Selective mGlu5 positive allosteric modulator; brain penetrant and orally bioavailable
Brand:  Tocris Bioscience™ 5693/10
Encompass
Product Code. 15747056
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
VU 0409551 is a selective mGlu5 positive allosteric modulator (EC50 = 260 nM). Selective for mGlu5 over other mGluR subtypes, as well as 66 other receptors and ion channels. Selectively potentiates mGlu5 signaling via Gαq. Exhibits wake-promoting, antipsychotic-like and cognition-enhancing effects in animal models. Brain penetrant and orally available.
Specifications
Specifications
[6,7-Dihydro-2-(phenoxymethyl)oxazolo[5,4-c]pyridin-5(4H)-yl](fluorophenyl)methanone
1363281-27-9
JNJ 46778212