missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ VU 6005649

Description
VU 6005649 is a positive allosteric modulator of mGlu7 and mGlu8 receptors (EC50 = 0.65 μM and 2.6 μM for mGlu7 and mGlu8, respectively). Displays >3.8 fold selectivity for mGlu7 and mGlu8 over other mGlu receptors (EC50 >10 μM for mGlu1,2,3,4,5,6). Also NK1 receptor antagonist (Ki= 650 nM, IC50 = 3.4 μM). Brain penetrant.
Specifications
Specifications
| Chemical Name or Material | 3-(2,3-Difluoro-4-methoxyphenyl)-2,5-dimethyl-7-(trifluoromethyl)pyrazolo[1,5-a]pyrimidine |
| CAS | 2137047-43-7 |
| Quantity | 50 mg |
| Molecular Formula | C16H12F5N3O |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?