missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWA5A Monoclonal Antibody (OTI3D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501569
This item is not returnable.
View return policy
Description
May play a role in tumorigenesis as a tumor suppressor. Altered expression of this protein and disruption of the molecular pathway it is involved in, may contribute directly to or modify tumorigenesis.Specifications
| VWA5A | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O00534 | |
| VWA5A | |
| Full length human recombit protein of human VWA5A produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D6 | |
| Unconjugated | |
| VWA5A | |
| BCSC-1, BCSC1, LOH11CR2A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4013 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction