missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWA5A Monoclonal Antibody (OTI3D6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501569S
This item is not returnable.
View return policy
Description
VWA5A Monoclonal antibody specifically detects VWA5A in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| VWA5A | |
| Monoclonal | |
| 0.69 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5TFE4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 221294 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D6 | |
| Unconjugated | |
| NT5DC1 | |
| 5830475I06Rik; AW552491; BCSC1; BCSC-1; breast cancer suppressor candidate 1; E130119J22; Loh11cr2a; loss of heterozygosity 11 chromosomal region 2 gene A protein; Loss of heterozygosity 11 chromosomal region 2 gene A protein homolog; loss of heterozygosity, 11, chromosomal region 2, gene A; loss of heterozygosity, 11, chromosomal region 2, gene A homolog; Masa1; masa-1; Mast cell surface antigen 1; mast cell surface antigen-1; ortholog of mouse AW551984; von Willebrand factor A domain containing 5A; von Willebrand factor A domain-containing protein 5A; VWA5A | |
| Full length human recombinant protein of human NT5DC1 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction