Learn More
VWA5A Mouse anti-Human, Clone: OTI2C6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501637
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May play a role in tumorigenesis as a tumor suppressor. Altered expression of this protein and disruption of the molecular pathway it is involved in, may contribute directly to or modify tumorigenesis.Specifications
| VWA5A | |
| Monoclonal | |
| Unconjugated | |
| VWA5A | |
| BCSC-1, BCSC1, LOH11CR2A | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4013 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2C6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O00534 | |
| VWA5A | |
| Full length human recombit protein of human VWA5A produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.