missing translation for 'onlineSavingsMsg'
Learn More
Learn More
vWF-A2 Antibody (7G2B11C10), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06526AF750
This item is not returnable.
View return policy
Description
vWF-A2 Monoclonal antibody specifically detects vWF-A2 in Human samples. It is validated for ELISA
Specifications
| vWF-A2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| 7G2B11C10 | |
| ELISA | |
| F8VWF, VWD, VWF von Willebrand factor | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human vWF-A2 (aa 1-2813; NP_000543.2). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction