missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ vWF-A2 Antibody (VWF/4458) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27026FR
This item is not returnable.
View return policy
Description
vWF-A2 Monoclonal antibody specifically detects vWF-A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| vWF-A2 | |
| Monoclonal | |
| DyLight 680 | |
| F8VWF, VWD, VWF von Willebrand factor | |
| Recombinant full-length human vWF-A2 (exact sequence is proprietary) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| VWF/4458 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 7450 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction