missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ vWF-A2 Antibody (VWF/4458), Novus Biologicals™
Mouse Monoclonal Antibody
€ 232.00 - € 462.00
Specifications
| Antigen | vWF-A2 |
|---|---|
| Clone | VWF/4458 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30043508
|
Novus Biologicals™
NBP3-26743-100ug |
100 μg |
€ 462.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30043870
|
Novus Biologicals™
NBP3-26743-20ug |
20 μg |
€ 232.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
vWF-A2 Monoclonal antibody specifically detects vWF-A2 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| vWF-A2 | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Human | |
| F8VWF, VWD, VWF von Willebrand factor | |
| Recombinant full-length human vWF-A2 (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| VWF/4458 | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7450 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title