missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ VWF Monoclonal Antibody (C1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543701
This item is not returnable.
View return policy
Description
VWF Monoclonal Antibody for Western Blot
Von Willebrand factor (vWf) is a series of self-aggregated structures, all derived from common glycoprotein subunits, synthesized in endothelial cells and megakaryocytes. The molecular mass of the monomeric vWf unit is 225 kDa. vWf binds to circulating factor VIIIc (antihemophilic factor), hereby stabilizing the latter thus being an important factor in the normal primary haemostasis.
Specifications
| VWF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P04275, Q62935 | |
| VWF | |
| Recombinant protein Von Willebrand Factor. The antigen corresponds to amino acid range 49-230 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG2a κ |
| Western Blot | |
| C1 | |
| Unconjugated | |
| VWF | |
| 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 116669, 7450 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction