missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VWF Monoclonal Antibody (OTI9F3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803609S
This item is not returnable.
View return policy
Description
VWF Monoclonal antibody specifically detects VWF in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| VWF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P04275 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 7450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI9F3 | |
| Unconjugated | |
| VWF | |
| 6820430P06Rik; AI551257; B130011O06Rik; C630030D09; Coagulation Factor VIII; coagulation factor VIII VWF; EMBL:CAB37852.1}; F8VWF; factor viii related antigen; von Willebrand antigen 2; von Willebrand antigen II; von Willebrand Disease (vWD); von Willebrand factor; Von Willebrand factor homolog; VWD; vwf; vwf {ECO:0000312 | |
| Human recombinant protein fragment corresponding to amino acids 764-1263 of human VWF produced in SF9 cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction