Learn More
VWF Mouse anti-Human, Clone: 907CT12.1.9, Abnova™
Mouse monoclonal antibody raised against human VWF.
Brand: Abnova MAB12355.400uL
Description
The glycoprotein encoded by this gene functions as both an antihemophilic factor carrier and a platelet-vessel wall mediator in the blood coagulation system. It is crucial to the hemostasis process. Mutations in this gene or deficiencies in this protein result in von Willebrand's disease. An unprocessed pseudogene has been found on chromosome 22. [provided by RefSeq]
Specifications
| VWF | |
| Monoclonal | |
| Unconjugated | |
| Flow Cytometry (1:100) Immunohistochemistry (1:25) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
| F8VWF/VWD | |
| Mouse | |
| Protein G purification | |
| RUO | |
| 7450 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Flow Cytometry, Immunohistochemistry (PFA fixed), Western Blot | |
| 907CT12.1.9 | |
| Mouse monoclonal antibody raised against human VWF. | |
| In PBS (0.09% sodium azide) | |
| VWF | |
| Recombinant protein corresponding to human VWF. | |
| 400 μL | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.