missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAC Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-82057AF700
This item is not returnable.
View return policy
Description
WAC Polyclonal antibody specifically detects WAC in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| WAC | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 1110067P07Rik, bA48B24, bA48B24.1, BM-016, FLJ31290, KIAA1844, MGC10753, PRO1741, WW domain containing adaptor with coiled-coil, WW domain-containing adapter protein with coiled-coil, WW domain-containing adaptor with coiled coil, Wwp4 | |
| WAC antibody was raised against a 19 amino acid peptide near the center of human WAC . The immunogen is located within amino acids 210 - 260 of WAC. Amino Acid Squence: CILSQTSRHNDRDYRLPRAE | |
| 0.1 mL | |
| Cell Biology | |
| 51322 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction