Learn More
asecos™ Wall Bracket for Roll with Cutting Edge
Product Code. 17886356 Shop All asecos Products
Click to view available options
Length (English):
19.69 in.
Unit Size:
Each
Product Code. 17886356
Brand: asecos™ 9905
This item requires a price update prior to being ordered.
Request a Price for this item here.
Wall Bracket for Roll with Cutting Edge provides safe and easy storage and separation of the roll.
  • Wall mounted bracket for Roll
  • With cutting edge for easy separation.
  • Dimensions exterior: W x D x H (mm) 500 x 230 x 270
Specifications
Material Steel
Type Spill Kit Dispenser
Length (English) 19.69 in.
Length (Metric) 500.00 mm
Width (English) 19.69 in.
Width (Metric) 500.00 mm
Height (English) 10.63 in.
Height (Metric) 270.00 mm
Weight (English) 6.31 lb.
Weight (Metric) 2.86 kg