Learn More
Bartels Rieger™ Wall Container
Product Code. 10519062
Change view
Click to view available options
Material:
ABS Plastic
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
10519062 ABS Plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10519062 Supplier Bartels Rieger™ Supplier No. 154812
  • ONLINE EXCLUSIVE PRICE
    € 48.40
    € 39.67 / Pack of 1
    Save € 8.73
    18% Off
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Depth (Metric) 80 mm
For Use With (Equipment) Eye Wash Bottle
Height (Metric) 320 mm
Includes Transparent Cover Marked with First-aid Symbol
Material ABS Plastic
Mounting Wall
Product Type Wall Container
Width (Metric) 180 mm