Learn More
Buerkle™ Wall-mount Draining Rack
Shop All Bürkle Products
Click to view available options
Positions:
33
77
Product Type:
With draining channel
Without draining channel
Height (Metric):
400 mm
500 mm
600 mm
Width (Metric):
400 mm
500 mm
600 mm
Unit Size:
Each
Description
A wall-mounted draining rack, including a wall mounting set (screws, dowels). A removable draining channel is optionally available, with a connection for a draining tube which can discharge directly into a laboratory wash basin.
- Draining rack PVC, rods polypropylene
- Including spanner for the rods
- Three different rod lengths to adapt to varying apparatus
- Shatterproof
- Rods click into place for secure positioning
Specifications
Specifications
| Includes | Spanner, wall mounting set of screws and dowels, box for the rods, five 60 mm rods, twenty six 100 mm rods, two 150 mm rods |
| Product Type | Without draining channel |
| Color | Blue/White |
| Material | PVC, Polypropylene |
| Positions | 33 |
| For Use With (Application) | Hygienic Drying of Apparatus |
| Size | 400 x 400 mm |
| Width (Metric) | 400 mm |
| Width (English) | 15.74 in. |
| Height (Metric) | 400 mm |
| Show More |