missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAPL Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-552IR
This item is not returnable.
View return policy
Description
WAPL Polyclonal antibody specifically detects WAPL in Human samples. It is validated for Western Blot
Specifications
| WAPL | |
| Polyclonal | |
| Western Blot | |
| FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
| The immunogen recognized by this antibody maps to a region between residues 1125 and 1175 of human wings apart like/KIAA0261 using the numbering given in TrEMBL entry Q7Z5K2 (GeneID 23063). | |
| 0.1 mL | |
| DNA Repair | |
| 23063 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 755 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction