missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WAPL Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 450.00 - € 708.00
Specifications
| Antigen | WAPL |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641454
|
Novus Biologicals
NBP3-21309-100ul |
100 μg |
€ 708.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677484
|
Novus Biologicals
NBP3-21309-25ul |
25 μg |
€ 450.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
WAPL Polyclonal antibody specifically detects WAPL in Human samples. It is validated for ImmunofluorescenceSpecifications
| WAPL | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair | |
| PBS, pH 7.2, 40% glycerol | |
| 23063 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| FOEKIAA0261friend of EBNA2 (Epstein-Barr virus nuclear protein 2), Friend of EBNA2 protein, WAPLwings apart-like protein homolog, wings apart-like homolog (Drosophila) | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: EEHEKNSHHIHKNADDSTKKPNAETTVASEIKETNDTWNSQFGKRPESPSEISPIKGSVRTGLFEWDNDFEDIRSEDCILSLDSDPLLEMKDD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title