Learn More
Brady™ Warning Sign without Text - Explosive Atmosphere
Product Code. 15561500 Shop All Brady Products
Change view
Click to view available options
Dimensions (L x W):
173 x 200 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
15561500 173 x 200 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15561500 Supplier Brady™ Supplier No. 255224
  • ONLINE EXCLUSIVE PRICE
    € 11.95
    € 10.77 / Pack of 1
    Save € 1.18
    9% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Warning sign for hazard identification.
  • Improves workplace safety
Specifications
Alternate Part Number PIC 313-TRI 200-B7525
Material Aluminum
Shape Triangular
Dimensions (L x W) 173 x 200 mm
Color Black on Yellow