missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WARS Polyclonal Antibody

WARS Polyclonal Antibody
Brand: Invitrogen PA576817
This item is not returnable.
View return policy
Description
WARS Polyclonal Antibody for Western Blot, ICC/IF
Specifications
| WARS | |
| Polyclonal | |
| Unconjugated | |
| WARS1 | |
| GAMMA-2; hWRS; IFI 53; IFI53; IFP 53; IFP53; Interferon-induced protein 53; T1-TrpRS; T2-TrpRS; TrpRS; tryptophan tRNA ligase 1, cytoplasmic; tryptophan--tRNA ligase, cytoplasmic; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, cytoplasmic; Wars; WARS protein; Wrs | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22375, 314442, 7453 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| P23381, P32921, Q6P7B0 | |
| WARS1 | |
| Recombinant full length Human WARS. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction