missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WARS2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543808
This item is not returnable.
View return policy
Description
Peptide sequence: TTKQKHDGTV GLLTYPVLQA ADILLYKSTH VPVGEDQVQH MELVQDLAQG Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 93%; Zebrafish: 93%.
Aminoacyl-tRNA synthetases catalyze the aminoacylation of tRNA by their cognate amino acid. Because of their central role in linking amino acids with nucleotide triplets contained in tRNAs, aminoacyl-tRNA synthetases are thought to be among the first proteins that appeared in evolution. Two forms of tryptophanyl-tRNA synthetase exist, a cytoplasmic form, named WARS, and a mitochondrial form, named WARS2. This gene encodes the mitochondrial tryptophanyl-tRNA synthetase. Two alternative transcripts encoding different isoforms have been described.
Specifications
| WARS2 | |
| Polyclonal | |
| Unconjugated | |
| WARS2 | |
| (Mt)TrpRS; 5730427B17Rik; 9430020O07Rik; AI413375; mitochondrial rryptophanyl-tRNA synthetase-like protein; mitochondrial tryptophanyl-tRNA synthetase 2; trpRS; tryptophan tRNA ligase 2, mitochondrial; tryptophan--tRNA ligase, mitochondrial; tryptophanyl tRNA synthetase 2 (mitochondrial); tryptophanyl tRNA synthetase 2, mitochondrial; Tryptophanyl-tRNA synthetase; tryptophanyl-tRNA synthetase, mitochondrial; tryptophanyl-tRNA synthetase, mitochondrial; tryptophan--tRNA ligase, mitochondrial; WARS2; zgc:110828 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10352 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UGM6 | |
| WARS2 | |
| Synthetic peptide directed towards the middle region of human WARS2. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction