missing translation for 'onlineSavingsMsg'
Learn More

WASF2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-12675-50ul

 View more versions of this product

Product Code. 18674557

  • € 183.00 / 50µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

WASF2 Polyclonal antibody specifically detects WASF2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

WASF2
Polyclonal
Western Blot 1:500-1:2000
IMD2, Protein WAVE-2, SCAR2suppressor of cyclic-AMP receptor (WASP-family), Verprolin homology domain-containing protein 2, WAS protein family, member 2, WASP family Verprolin-homologous protein 2, WAVE2dJ393P12.2, wiskWASP family protein member 2
Produced in rabbits immunized with E. coli-derived Human WASF2 fragment
50 μg
Cancer, Cell Biology, Cytoskeleton Markers, Immunology, Signal Transduction
10163
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS, pH7.0
Rabbit
Antigen and protein A Affinity-purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.