missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASHC4 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44291AF405
This item is not returnable.
View return policy
Description
WASHC4 Polyclonal antibody specifically detects WASHC4 in Human, Mouse samples. It is validated for Western Blot
Specifications
| WASHC4 | |
| Polyclonal | |
| Western Blot | |
| KIAA1033, Strumpellin And WASH-Interacting Protein, SWIP, WASH Complex Subunit 7 | |
| The epitope recognized by this antibody maps to a region between residue 1123 to 1173 of human WASH complex subunit 7 using the numbering given in entry Q2M389.2 (GeneID 23325). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23325 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction