missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WASHC4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-44291CL1
This item is not returnable.
View return policy
Description
WASHC4 Polyclonal antibody specifically detects WASHC4 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| WASHC4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 23325 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| KIAA1033, Strumpellin And WASH-Interacting Protein, SWIP, WASH Complex Subunit 7 | |
| The epitope recognized by this antibody maps to a region between residue 1123 to 1173 of human WASH complex subunit 7 using the numbering given in entry Q2M389.2 (GeneID 23325). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction